Anti-MARCH7 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP16616AEA 348 EUR
ABGEAP16616A
Anti-MARCH7 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MARCH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR