Anti-CDKN1B Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP3874AEA 348 EUR
ABGEAP3874A
Anti-CDKN1B Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR