Alumiinioksidi 50% in water, NanoArc® X1121W

Toimittaja: Thermo Scientific

Synonyms: Alumina C

44925.22EA 73.3 EUR
44925.22
Alumiinioksidi 50% in water, NanoArc® X1121W
Alumiinioksidi

Applications: Aluminum oxide water borne dispersion suitable for incorporation in water compatible coatings

Kaava: Al₂O₃
MW: 101,96 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003424
CAS-numero: 1344-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR