Amberlite® FPC3500 (H) ion exchange resin
Toimittaja: Thermo Scientific
45903.A1EA 297 EUR
45903.A1 45903.30
Amberlite® FPC3500 (H) ion exchange resin
Amberlite® FPC3500 (H)
Applications: Suitable for the neutralization of strong bases, the recovery of metallic ions, and the isolation and concentration of antibiotics, basic amino acids, enzymes, and peptides.