Platina 3% on carbon paste B103032-3

Toimittaja: Thermo Scientific

H36301.14EA 221 EUR
H36301.14 H36301.06 H36301.22
Platina 3% on carbon paste B103032-3
Platina
Kaava: Pt
MW: 195,08 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4
EINECS: 231-116-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR