2-(4-Methylphenyl)benzimidazole 97%

Varoitus

H58894.04EA 452 EUR
H58894.04
2-(4-Methylphenyl)benzimidazole 97%
2-(4-Methylphenyl)benzimidazole
Kaava: C₁₄H₁₂N₂
MW: 208,26 g/mol
Sulamispiste: 265…269 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00209485
CAS-numero: 120-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR