Z-Arg(NO2)-OH 98%

Toimittaja: Thermo Scientific

H63392.14EA 408 EUR
H63392.14 H63392.06
Z-Arg(NO2)-OH 98%
Z-Arg(NO2)-OH
Kaava: C₁₄H₁₉N₅O₆
MDL numero: MFCD00038092
CAS-numero: 2304-98-5
EINECS: 219-970-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR