3-Bromo-4-nitrobenzotrifluoride 95%

Varoitus

H64376.MDEA 57 EUR
H64376.MD
3-Bromo-4-nitrobenzotrifluoride 95%
3-Bromo-4-nitrobenzotrifluoride
Kaava: C₇H₃BrF₃NO₂
MW: 270,01 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09998160
CAS-numero: 132839-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR