N-Benzyloxycarbonyl-L-aspartic acid-4-tert-butyl ester-1-methyl ester 98%

Toimittaja: Thermo Scientific

H66384.06EA 151 EUR
H66384.06 H66384.14
N-Benzyloxycarbonyl-L-aspartic acid-4-tert-butyl ester-1-methyl ester 98%
N-Benzyloxycarbonyl-L-aspartic acid-4-tert-butyl ester-1-methyl ester
Kaava: C₁₇H₂₃NO₆
MDL numero: MFCD00270581
CAS-numero: 63327-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR