Wash buffer, 0,5% Tween 20, pH 8,0

Toimittaja: Thermo Fisher Scientific
J60165.APEA 108 EUR
ALFAJ60165.AP
Wash buffer, 0,5% Tween 20, pH 8,0
Physical Form: Liquid
Odour: Characteristic
Applications: For His-tag protein purification
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR