Pig Insulin (from Pancreas)

Toimittaja: Thermo Fisher Scientific
J61321.MDEA 1670 EUR
ALFAJ61321.MD
Pig Insulin (from Pancreas)
Purity: 97%
CAS No.: 12584-58-6
Molecular Formula: C256H381N65O76S6
Formula Weight: 5777,59
Storage Temperature: −30 to −10 °C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00131380

Applications: A hormone that is central to regulating energy and glucose metabolism in the body
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR