Calpain inhibitor 3

Toimittaja: Thermo Fisher Scientific
J62919.MBEA 359 EUR
J62919.MB
Calpain inhibitor 3
Proteiinit
Purity: 95+%
CAS No.: 88191-84-8
Molecular Formula: C19H35N3O4S
Formula Weight: 401.60
Storage Temperature: 2 to 8 °C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO at 25 mg/ml
MDL No.: MFCD00798796

Applications: Inhibits calpain proteolysis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR