Casein blocking buffer, 3% in TBS, with 0,02% sodium azide
Toimittaja: Thermo Fisher Scientific
J62935.APEA 189 EUR
J62935.AP J62935.AK
Casein blocking buffer, 3% in TBS, with 0,02% sodium azide
Biokemialliset puskuriliuokset Elektroforeesipuskurit Blokkauspuskurit
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
MDL No.: MFCD01634587
Applications: For Western Blots and ELISA