Bradykinin

Toimittaja: Thermo Fisher Scientific
J63131.MAEA 179 EUR
J63131.MA
Bradykinin
Proteiinit
An endogenous vasodilator peptide

CAS No.: 58-82-2
Molecular Formula: C50H73N15O11
Formula Weight: 1060,23
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00076258
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR