LB medium, Miller
Toimittaja: Thermo Fisher Scientific
J63905.K2EA 237 EUR
ALFAJ63905.K2
LB medium, Miller
Used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates. Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins.
Physical Form: Liquid