trans-4,4'-Diphenylstilbene ≥99%

L07837.MDEA 70.3 EUR
L07837.MD L07837.03
trans-4,4'-Diphenylstilbene ≥99%
trans-4,4'-Diphenylstilbene
Kaava: C₂₆H₂₀
Sulamispiste: 308…310 °C (haj. lämpötila)
MDL numero: MFCD00003063
CAS-numero: 2039-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR