Binding and elution buffers for antibody purification, Pierce
Toimittaja: Thermo Scientific
These buffers ensure high-yield, non denaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads.
  • Ready to use
  • Optimised for protein A, protein G and protein A/G
TILAUS
Laajenna kaikki / Pienennä kaikki
Tuote Pak. VWR-tuoteluettelon nro Määrä
Protein A IgG binding buffer 1 l PIER21001
Protein A IgG binding buffer 3,75 l PIER21007
Protein G IgG binding buffer 1 l PIER21019
Protein G IgG binding buffer 3,75 l PIER21011
IgG elution buffer 1 l PIER21004
IgG elution buffer 3,75 l PIER21009
IgG elution buffer, pH 2,0 1 l PIER21028
IgM elution buffer 500 ml PIER21017
Gentle Ag/Ab elution buffer, pH 6,6 500 ml PIER21027
Gentle Ag/Ab elution buffer, pH 6,6 3,75 l PIER21013
Gentle Ag/Ab binding buffer, pH 8,0 1 l PIER21020
Gentle Ag/Ab binding buffer, pH 8,0 3,75 l PIER21012
Gentle Ag/Ab binding and elution buffer kit 100 ml PIER21030
Protein A/G IgG Binding Buffer 240 ml PIER54200
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Syötä tarkemmat tiedot.
Valitse tuote johon etsit Sertifikaattia.
Ilmoita eränumero. Tämä kenttä on pakollinen. Jos et tiedä VWR Chemicalsin eränumeroa syötä "x".
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
tai
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Kuvaus Tuoteluettelon nro Availability Tuote Sinun hintasi Price Per lkm