Thiol-reactive dyes and reagents
Toimittaja: Biotium
91052.EA 138 EUR
91052. 91030. 91042. 91020. 91046. 91024. 91010. 91032. 91022. 91044. 91038. 91028. 91036. 91048. 91026. 91029. 92015. 92013. 91053. 91031. 91045. 91023. 91035. 91043. 91021. 91011. 91055. 91033. 91027. 91025. 91037.
Thiol-reactive dyes and reagents
Proteiinien yhdistämis- ja leimausjärjestelmät
Reactive dyes for labeling cysteine residues in proteins and other thiol molecules. For monitoring and validating the sterilisation process.
  • Soluble in water, DMSO or DMF
  • Wide range of bioindicators for:
  • Steam
  • Ethylene oxide
  • Dry heat
  • Hydrogen peroxide
  • Irradiation
Available in ampoule, self-contained, strip, suspension and coupon formats in several materials such as glass, aluminium, poliflex, steel and PVC.
All biological indicators have a certificate reporting D and Z values, survival time, kill time, population and purity.
Sertifikaatit: Conform in accordance with ISO 11138 and EN 866 regulations.