(+)-2-Methyl-L-serine 99%

Toimittaja: Thermo Scientific

ACRO290012500EA 231 EUR
ACRO290012500
(+)-2-Methyl-L-serine 99%
(+)-2-Methyl-L-serine

Appearance: White Powder

Kaava: C₄H₉NO₃
Sulamispiste: 286…288 °C
Storage Temperature: Huoneenlämpö
CAS-numero: 16820-18-1

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
TLC ≥98.5 %
Specific optical rotation +2.3° to +4.3° (25°C, 589 nm) (c=5, 6 N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR