Dowex® 50WX2 (H) 200-400 mesh ion-exchange resin
|
Vaara
|
ACRO203045000EA
237
EUR
ACRO203045000
ACRO203041000
Dowex® 50WX2 (H) 200-400 mesh ion-exchange resin
Dowex® 50WX2 (H)
Storage Temperature:
Huoneenlämpö |
MDL numero:
MFCD00132722 CAS-numero: 12612-37-2 |
Testitulosten erittely
Appearance | Beads |
Particle size | ≤15 % On 70 mesh (cumulative) |
Particle size | ≤15 % Through 270 mesh |
Wet volume capacity | ≥0.6 meq/ml H form |
Water retention capacity | 74 to 82 % H form |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...