Tris[2-(diphenylphosphino)ethyl]phosphine 97%

Varoitus

ACRO316830050EA 450 EUR
ACRO316830050 ACRO316830010 ACRO316830250
Tris[2-(diphenylphosphino)ethyl]phosphine 97%
Tris[2-(diphenylphosphino)ethyl]phosphine
Kaava: C₄₂H₄₂P₄
MW: 670,693 g/mol
Sulamispiste: 134…139 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010009
CAS-numero: 23582-03-8
UN: 3464
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
Melting point 134 to 139 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR