(R)-β-Phenylalaninol 95%, 98% ee

Vaara

ACRO363120010EA 228 EUR
ACRO363120010
(R)-β-Phenylalaninol 95%, 98% ee
(R)-β-Phenylalaninol
Kaava: C₉H₁₃NO
MW: 151,2056 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01311791
CAS-numero: 170564-98-4
UN: 3259
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White to light yellow Low melting solid or viscous liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR