Sign and label printer, BBP™30
Toimittaja: Brady
116-0613EA 1420 EUR
116-0613 116-0612
Sign and label printer, BBP™30
Tulostimet ja tulostimien lisävarusteet Label Printers
The BBP™30 sign and label printer offers trouble-free printing and a wide range of durable signs for a small investment. With the BBP30 peripheral printer system every laboratory has the potential to create durable signs that reduce risk and accidents or increase productivity and compliance, all for a small investment. Maximum 600 labels (2×4", 50×100 mm) or 300 larger labels per day.
  • Single colour printing, 300 dpi, on continuous or die-cut labels with built-in cutter
  • Works with easy to use ‘drop-in’ label and ink ribbon cartridges for easy printing
  • Printer automatically recognises the cartridge and will auto-calibrate to optimal settings without any difficulties
  • Wide range of label types and materials available
Sertifikaatit: UL, cUL, CE, WEEE, RoHS, RCM (Australia / New Zealand), EAC (Russia, Belarus & Kazakhstan), CCC (China) and China RoHS
Toimitustiedot: Supplied with power cord, USB cable, cleaning kit, cutter cleaning tool and quick start guide.