VWR® RNA Loading Buffer (2X)
Toimittaja: Avantor
VWR®
1B1607-1MLEA 0 EUR
1B1607-1ML 1B1606-1ML
VWR® RNA Loading Buffer (2X)
Biokemialliset puskuriliuokset Elektroforeesipuskurit Loading Buffers
RNA loading buffers (2X) are premixed loading dyes for the preparation of RNA samples for agarose or polyacrylamide electrophoresis.
  • With ethidium bromide, 2X provides immediate post-electrophoresis visualization of the RNA
  • Efficient RNA separation
  • Contains formamide as a denaturing agent and RNA stabilizer, as well as bromophenol blue and xylene cyanol FF tracking dyes for monitoring of RNA migration