trans-4-Heksen-1-oli predominantly trans 97%

Toimittaja: Thermo Scientific
Varoitus

ACRO217950050EA 77.9 EUR
ACRO217950050 ACRO217950250
trans-4-Heksen-1-oli predominantly trans 97%
trans-4-Heksen-1-oli
Kaava: C₆H₁₂O
MW: 100,16 g/mol
Kiehumispiste: 159…160 °C (760 mmHg)
Tiheys: 0,85 g/cm³
Leimahduspiste: 61 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009713
CAS-numero: 928-92-7

Order Now

Testitulosten erittely

Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR