1H-Indazole-4-carboxylic acid 97%
Toimittaja: Thermo Scientific
|
Varoitus
|
ACRO430090010EA
177
EUR
ACRO430090010
1H-Indazole-4-carboxylic acid 97%
1H-Indazole-4-carboxylic acid
Kaava:
C₈H₆N₂O₂ MW: 162,15 g/mol Sulamispiste: 209,5…211 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD03840644 CAS-numero: 677306-38-6 |
Testitulosten erittely
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 207 to 212 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...