2-Aminopyrimidin-5-ol 95%
|
Varoitus
|
ACRO434330050EA
561
EUR
ACRO434330050
ACRO434330010
2-Aminopyrimidin-5-ol 95%
2-Aminopyrimidin-5-ol
Kaava:
C₄H₅N₃O MW: 111,1 g/mol Sulamispiste: 198,6…199 °C (haj. lämpötila) Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD08062411 CAS-numero: 143489-45-6 |
Testitulosten erittely
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
HPLC | ≥98.5 % |
Specific optical rotation | -6° to -9° (20°C, 589 nm) (C=0.5%, Ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...