3-Amino-6-bromo-1H-indazole 95%

Toimittaja: Thermo Scientific
Varoitus

ACRO435780050EA 254 EUR
ACRO435780050
3-Amino-6-bromo-1H-indazole 95%
3-Amino-6-bromo-1H-indazole
Kaava: C₇H₆BrN₃
Sulamispiste: 175…178 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05665872
CAS-numero: 404827-77-6
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 235 to 240 °C
HPLC ≥94.0 %
Specific optical rotation +40° to +44° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR