L-Methionine sulphone 98%

Toimittaja: Thermo Scientific

ACRO439000050EA 124 EUR
ACRO439000050 ACRO439000010
L-Methionine sulphone 98%
L-Methionine sulphone
Kaava: C₅H₁₁NO₄S
Sulamispiste: 242,3…243.1 °C (haj. lämpötila)
Storage Temperature: Jääkaappi
MDL numero: MFCD00066020
CAS-numero: 7314-32-1

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
TLC ≥97.5 %
Elemental Analysis 32.31 to 33.97 % C
Elemental Analysis 7.54 to 7.92 % N
Specific optical rotation +11.7° to +13.7° (24°C, 589 nm) (c=2, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR