2-Amino-3-iodopyridine 97%

Toimittaja: Thermo Scientific
Varoitus

ACRO441480050EA 365 EUR
ACRO441480050
2-Amino-3-iodopyridine 97%
2-Amino-3-iodopyridine
Kaava: C₅H₅IN₂
MW: 220,01 g/mol
Sulamispiste: 89,6…90.6 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01651885
CAS-numero: 104830-06-0

Order Now

Testitulosten erittely

Appearance White to light brown Powder or needles
Infrared spectrum Conforms
Melting point 86 to 92 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR