4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%

Toimittaja: Thermo Scientific
Varoitus

ACRO445000050EA 691 EUR
ACRO445000050
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester
Kaava: C₁₆H₂₃BO₄
MW: 290,1624 g/mol
Sulamispiste: 41…43 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD08704684
CAS-numero: 859169-20-3

Order Now

Testitulosten erittely

Appearance White to off-white Low melting solid
Infrared spectrum Conforms
Melting point 38 to 46 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR