ESD safety ankle boots, lace-up, New Classic, Leroy S2
Toimittaja: Elten
ELTN76628/39EA 80.9 EUR
ELTN76628/39 ELTN76628/38 ELTN76628/40 ELTN76628/44 ELTN76628/43 ELTN76628/42 ELTN76628/41 ELTN76628/48 ELTN76628/47 ELTN76628/46 ELTN76628/45
ESD safety ankle boots, lace-up, New Classic, Leroy S2
ESD safety ankle boots made from cowhide leather, a full padded bellows tongue and a dual-density PU/PU sole New Classics.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Scuff cap protects the leather upper
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
Sertifikaatit: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC