These compressor-cooled incubators, with forced air convection and fan speed control, feature an acid-proof stainless steel (DIN1.4301) inner chamber and a powder coated sheet exterior. The adjustable heating and cooling system provides a temperature range between –10 to +100 °C. All models have a solid outer door and an internal glass door. The larger IL 400CR and IL 750CR models have double outer doors. All models are equipped with levelling castors, 30 mm access port, internal light, USB port to allow direct recording and data transfer onto a flash drive and Ethernet (LAN) port for remote control.
- Multi-functional microprocessor controller with colour LCD touch screen, displays values in tabular or graphic form
- Access control via login, 20 user-program memory, 7 day programming
- Administrator function to manage user accounts
- Multi-segment temperature/time profile (up to 100) with adjustable ramps
- Real time clock displays date/time/weekday
- Adjustable start delay feature
- Adjustable over- and under-temperature safety thermostat Class 3.3 protects samples and the incubator
Safety features include audible and visible temperature alarms, power failure control system, audible clock alarm, automatic defrosting function, door lock and open door alarm, temperature sensor fail alarm and auto-diagnostic function.
Units feature Premium control software and aid GLP compliance with password protection settings, events registry and an internal memory which stores up to 4100 data records for each user, allowing the values to be viewed on the display or a PC in tabular or graphic form.
Lisävarusteiden tiedot: The standard spare shelves have a maximum workload of 25 kg per shelf. The reinforced stainless steel shelves (optional for IL 240CR and IL 400CR) have a maximum workload of 100 kg per shelf. Please note the maximum chamber capacity as shown in the specification.
Toimitustiedot: All models are supplied with test certificate (data measured at 37 °C), Ethernet cable and Premium Control Software on USB stick. The IL 750CR models are supplied with three reinforced stainless steel wire shelves, the IL 240CR and IL 400CR models are supplied with three standard stainless steel wire shelves.