The Clino® CR4 EM-GMP trolley system made of stainless steel suitable for cleanrooms and for high sensitive areas that require minimal space.
- Individually configurable through Clino modular design system
- EasyMop GMP with system box certified for areas under sterille conditions
- Highest efficiency and process reliability in single step technique
- Including contact-free dropping device MopDrop
- Basic module mini with high end finish and castors (100 mm)
The trolley is an high-end solution for the cleanroom cleaning in low classified cleanrooms up to sterile areas. The EasyMop pre-preparation systems allows a validatable and reproducible cleaning and disinfection of surfaces.
Sertifikaatit: With IPA certificate.
Toimitustiedot: Supplied with 2×system boxes EasyMop GMP, 2×4-litre stainless steel container, disposal unit (1×35 L) and stainless steel container (9 L).