Cleaning trolley Clino®, CR4 FP-GMP
Toimittaja: Pfennig
PPSA3540100EA 2755.44 EUR
PPSA3540100
Cleaning trolley Clino®, CR4 FP-GMP
The Clino® CR4 FP-GMP trolley system made of stainless steel suitable for cleanrooms and for high sensitive areas that require minimal space. ClinoLink cleanroom suitable connection system for individual attachment of accessory parts.
  • Individually configurable through Clino modular design system
  • Certified flat wringer ringo GMP for sterile areas
  • Full flexibility in single- and two-step technique
  • Basic module mini with high end finish and castors (100 mm)
  • Includes contact-free dropping device MopDrop
Sertifikaatit: With IPA certificate.
Toimitustiedot: Supplied with stainless steel container (25 and 9 L) and disposal unit (1×35 L).