3-in-1 pH/ATC probe, Orion™ 9109WP Triode™
Toimittaja: Thermo Fisher Scientific
662-1146EA 467 EUR
ORIO9109WP ORIO9109WL
3-in-1 pH/ATC probe, Orion™ 9109WP Triode™
This highly-versatile 3-in-1 pH/ATC probe can be used in either the field or a plant.
  • Contains a low-maintenance, gel-filled electrode with built-in ATC
  • Has an Ag/AgCl internal reference system, sealed reference
Can be used with Orion 260A/A+, 261, 261S, 265A/A+, 266 and 266S meters.