Anti-FTL Goat Polyclonal Antibody (Biotinylated)

Toimittaja: Everest Biotechnology
EVBIEB09092-BEA 325.5 EUR
EVBIEB09092-B
Anti-FTL Goat Polyclonal Antibody (Biotinylated)
Vasta-aineet
Type: Primary
Antigen: FTL
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR