Signs, floor signs, DuraStripe®, Mean Lean, corners
Toimittaja: Ergomat
ERGO4070059COREEA 61.6 EUR
ERGO4070059CORE ERGO4070059COGN ERGO4070059COBL
Signs, floor signs, DuraStripe®, Mean Lean, corners
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe program makes it the only total floor marking solution available on the market today. Economic than painting the floors.
  • Ideal for less than perfect floors
  • Simple and easy peel-&-stick installation
  • No dry time, cure time, fumes or messy clean up to limit productivity
Mean Lean has a durable, smooth surface layer that cleans easily and resists damage from pivoting tow-motor wheels. Because of its low profile, Mean Lean does not have a beveled edge. It is available in 51, 76 and 102 mm standard widths and 7 Standard colors.