Anti-ITGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Toimittaja: Bioss
BOSSBS-0486R-A647EA 866 EUR
BOSSBS-0486R-A647
Anti-ITGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
Type: Primary
Antigen: ITGB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR