Anti-BNIP3L Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP1320AEA 360 EUR
ABGEAP1320A
Anti-BNIP3L Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: BNIP3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 1–100 aa
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR