Anti-EZH2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP2512CEA 360 EUR
ABGEAP2512C
Anti-EZH2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: EZH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR