Anti-P2RX5 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP14059BEA 348 EUR
ABGEAP14059B
Anti-P2RX5 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: P2RX5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR