Serological pipettes
Toimittaja: Hecht
631-9323EA 5.5 EUR
631-9323
Serological pipettes
Pipetit ja pipetintäyttäjät Serologiset pipetit
Serological pipettes made of clear glass.
  • Total delivery (calibrated up to tip)
  • Zero at tip
  • Short line graduation
  • Fused amber graduation “ASSISTENT-PERMANENT”
  • Tolerance 1,5 times the official tolerance of class AS
Pakkaus: Individually plastic wrapped