Powered air respirator (PAPR) system, Proflow SC
Toimittaja: Scott Safety
SCTS2029808EA 1110 EUR
SCTS2029808
Powered air respirator (PAPR) system, Proflow SC
Hengityssuojaimet Hengityssuojajärjestelmät
Proflow SC is a lightweight and compact system that incorporates a curved backplate ensuring user comfort even for long periods. Electronic control compensates for higher filter resistance to ensure the correct flow is maintained.
  • Lightweight NiMH battery provides full shift operation
  • Simple recharging system, recharging takes only 6 hours
  • On board chip data logs downloadable usage data to provide full service and maintenance information
Sertifikaatit: EN 12941/2