Dr Ehrenstorfer standard, Benomyl, EPA 627, EPA 627 and 631

Toimittaja: LGC Standards
Dr Ehrenstorfer
EHERC10490000EA 148 EUR
EHERC10490000
Dr Ehrenstorfer standard, Benomyl, EPA 627, EPA 627 and 631
Standardiaineet ja vertailumateriaalit Organic Standards
  • Form: Powder
  • CAS.No: 17804-35-2
  • Melting point: 140 °C
  • Density: 0,38 g/cm³
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR