J.T.Baker® BAKER INSTRA-ANALYZED®, Zinc Standard Solution, 1000 µg/L Zn, reagent for AAS
Toimittaja: Avantor
J.T.Baker® BAKER INSTRA-ANALYZED®
6827.0100EA 77.69 EUR
6827.0100
J.T.Baker® BAKER INSTRA-ANALYZED®, Zinc Standard Solution, 1000 µg/L Zn, reagent for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Zinc Standards
Zinc standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Sertifikaatit: Traceable to NIST.