J.T.Baker® BAKERBOND spe™ PAH Soil, SPE Columns for Special Applications
Toimittaja: Avantor
J.T.Baker® BAKERBOND spe™
7518-08EA 380 EUR
7518-08
J.T.Baker® BAKERBOND spe™ PAH Soil, SPE Columns for Special Applications
Kromatografiakolonnit
This solid phase extraction column is designed for the extraction and clean-up of Poly Aromatic Hydrocarbons (PAH) from soil, including the 16 priority pollutant EPA PAH’s and for several applications using different SPE columns.