J.T.Baker® BAKERBOND spe™ PCB-N, SPE Columns for Special Applications
Toimittaja: Avantor
J.T.Baker® BAKERBOND spe™
7524-04EA 380 EUR
7524-04
J.T.Baker® BAKERBOND spe™ PCB-N, SPE Columns for Special Applications
Kromatografiakolonnit
This solid phase extraction column is designed for the extraction of PCB’s in oil.