Anti-F8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BOSSBS-0332R-A488EA 866 EUR
BOSSBS-0332R-A488
Anti-F8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Factor VIII, along with calcium and phospholipid, acts as a cofactor for factor IXa when it converts factor X to the activated form, factor Xa.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: factor VIII/FVIII
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR