Anti-CD27 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BOSSBS-2491R-A488EA 858 EUR
BOSSBS-2491R-A488
Anti-CD27 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Receptor for CD70/CD27L. May play a role in survival of activated T-cells. May play a role in apoptosis through association with SIVA1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD27
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR